siRNA Details
Gene Symbol: Krt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008473.2 | NP_032499.2 | 3 | 904 | ||
GenBank mRNA | AK019521.1 | - | 3 | 905 | ||
BC117842.1 | - | 3 | 871 | 40093577 | ||
BC117843.1 | - | 3 | 871 | 40093580 | ||
M10937.1 | - | 3 | 912 |

More Information
Other Species Targeted

Gene Ontology Categories:
