siRNA Details
Gene Symbol: Ly6e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164036.1 | NP_001157508.1 | 4 | 1400 | ||
NM_001164037.1 | NP_001157509.1 | 5 | 1273 | |||
NM_001164038.1 | NP_001157510.1 | 5 | 1189 | |||
NM_001164039.1 | NP_001157511.1 | 5 | 1186 | |||
NM_001164040.1 | NP_001157512.1 | 4 | 983 | |||
NM_008529.3 | NP_032555.1 | 4 | 1397 | |||
GenBank mRNA | AK014207.1 | - | 4 | 854 | ||
BC002116.1 | - | 4 | 800 | 3490293 | ||
BC003926.1 | - | 4 | 803 | 3485382 | ||
BC005684.1 | - | 4 | 835 | 3155116 | ||
BC037143.1 | - | 3 | 915 | |||
BC040691.1 | - | 4 | 820 | 5025135 | ||
BC053523.1 | - | 4 | 803 | 4205673 | ||
BC080711.1 | - | 4 | 800 | 3969173 | ||
U04268.1 | - | 3 | 765 | |||
U09192.1 | - | 4 | 790 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
