siRNA Details
Gene Symbol: Mobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039364.2 | NP_001034453.1 | 3 | 304 | ||
NM_008614.2 | NP_032640.1 | 4 | 383 | |||
GenBank mRNA | AK013799.1 | - | 4 | 384 | ||
AK028057.1 | - | 4 | 383 | |||
AK083103.1 | - | 4 | 384 | |||
BY257495.1 | - | 3 | 305 | |||
U81317.1 | - | 3 | 303 |
