siRNA Details
Gene Symbol: TEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000459.4 | NP_000450.2 | 23 | 4448 | ||
NM_001290077.1 | NP_001277006.1 | 22 | 4319 | |||
NM_001290078.1 | NP_001277007.1 | 21 | 4004 | |||
XM_005251561.1 | XP_005251618.1 | 23 | 4112 | |||
XM_005251563.1 | XP_005251620.1 | 22 | 3983 | |||
GenBank mRNA | AB208796.1 | - | 23 | 4126 | ||
AW020953.1 | - | 1 | 272 | |||
BC035514.1 | - | 23 | 4123 | 5228999 | ||
L06139.1 | - | 24 | 3815 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
