siRNA Details
Gene Symbol: Naip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008670.2 | NP_032696.2 | 5 | 844 | ||
GenBank mRNA | AF007769.1 | - | 4 | 941 | ||
AF135491.1 | - | 5 | 844 | |||
BC047215.1 | - | 5 | 849 | |||
BC132413.1 | - | 4 | 891 | 40130690 | ||
BC145251.1 | - | 4 | 891 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
