siRNA Details
Gene Symbol: Map2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039934.1 | NP_001035023.1 | 7 | 733 | ||
NM_008632.2 | NP_032658.2 | 7 | 733 | |||
GenBank mRNA | AK039532.1 | - | 2 | 109 | ||
AK079618.1 | - | 5 | 762 | |||
AK086484.1 | - | 5 | 597 | |||
AK141458.1 | - | 5 | 498 | |||
AK162828.1 | - | 2 | 96 | |||
AK164828.1 | - | 2 | 183 | |||
AK166380.1 | - | 7 | 749 | |||
BC048781.1 | - | 3 | 431 | |||
BC051410.1 | - | 3 | 428 | |||
BC052446.1 | - | 7 | 733 | |||
BC150945.1 | - | 7 | 732 | 9087856 | ||
CN538992.1 | - | 3 | 378 | |||
M21041.1 | - | 5 | 499 |
Gene Symbol: A730034C02 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042880.1 | - | 2 | 142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
