siRNA Details
Gene Symbol: AKT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014431.1 | NP_001014431.1 | 6 | 837 | ||
NM_001014432.1 | NP_001014432.1 | 7 | 921 | |||
NM_005163.2 | NP_005154.2 | 6 | 1051 | |||
XM_005267401.1 | XP_005267458.1 | 6 | 986 | |||
XM_017021075.1 | XP_016876564.1 | 7 | 1164 | |||
XM_017021076.1 | XP_016876565.1 | 7 | 1070 | |||
XM_017021077.1 | XP_016876566.1 | 6 | 986 | |||
XM_017021078.1 | XP_016876567.1 | 7 | 697 | |||
GenBank mRNA | AB451242.1 | - | 5 | 497 | ||
AB451367.1 | - | 5 | 497 | |||
AK122894.1 | - | 6 | 1978 | |||
AK299310.1 | - | 4 | 441 | |||
AK314256.1 | - | 6 | 650 | |||
BC000479.2 | - | 6 | 772 | 2964603 | ||
BC084538.1 | - | 7 | 915 | 6645486 | ||
BX647722.1 | - | 1 | 3426 | |||
BX648205.1 | - | 5 | 1455 | |||
KU177898.1 | - | 5 | 497 | |||
M63167.1 | - | 6 | 695 | |||
X61037.1 | - | 3 | 311 |

More Information
Other Species Targeted

Gene Ontology Categories:
