siRNA Details
Gene Symbol: Myo1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053214.2 | NP_444444.2 | 7 | 708 | ||
GenBank mRNA | AK153908.1 | - | 7 | 671 | ||
AK158333.1 | - | 7 | 623 | |||
BC046502.1 | - | 7 | 696 | 4167558 | ||
X97650.1 | - | 7 | 626 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
