siRNA Details
Gene Symbol: Mdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135162.1 | - | 9 | 1288 | ||
AK160553.1 | - | 9 | 1258 | |||
AK167809.1 | - | 9 | 1258 | |||
M16229.1 | - | 9 | 1234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
