siRNA Details
Gene Symbol: Nrap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286552.1 | NP_001273481.1 | 2, 3 | 327 | ||
NM_008733.4 | NP_032759.3 | 3, 4 | 422 | |||
NM_198059.3 | NP_932307.2 | 3, 4 | 422 | |||
GenBank mRNA | AK138925.1 | - | 2, 3 | 293 | ||
AY177623.1 | - | 3, 4 | 419 | |||
BC145109.1 | - | 2, 3 | 234 | |||
U76618.1 | - | 1783 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
