siRNA Details
Gene Symbol: Nsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008739.3 | NP_032765.3 | 2 | 203 | ||
GenBank mRNA | AK049663.1 | - | 2 | 159 | ||
AK145783.1 | - | 2 | 204 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107337, XM_008771494, XM_006253627, XM_006253620, XM_006253621, and others.
Gene Ontology Categories:
