siRNA Details
Gene Symbol: Nsf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008740.4 | NP_032766.2 | 4, 5 | 311 | ||
GenBank mRNA | AK042341.1 | - | 5, 6 | 329 | ||
AK049281.1 | - | 4, 5 | 281 | |||
AK051430.1 | - | 4, 5 | 280 | |||
AK085086.1 | - | 4, 5 | 265 | |||
AK141986.1 | - | 4, 5 | 280 | |||
AK153905.1 | - | 4, 5 | 280 | |||
BC006627.1 | - | 4, 5 | 316 | 3968733 | ||
BC019167.1 | - | 4, 5 | 264 | 4506351 | ||
U10120.1 | - | 4, 5 | 269 |
