siRNA Details
Gene Symbol: Orc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271526.1 | NP_001258455.1 | 3, 4 | 493 | ||
NM_008765.3 | NP_032791.1 | 3, 4 | 499 | |||
GenBank mRNA | AK048408.1 | - | 3, 4 | 318 | ||
AK051747.1 | - | 3, 4 | 328 | |||
AK133604.1 | - | 3, 4 | 318 | |||
AK146281.1 | - | 3, 4 | 318 | |||
AK146859.1 | - | 3, 4 | 312 | |||
BC015257.1 | - | 3, 4 | 309 | 4234871 | ||
U27457.1 | - | 3, 4 | 318 |

More Information
Other Species Targeted

Gene Ontology Categories:
