siRNA Details
Gene Symbol: Nfic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008688.3 | NP_032714.1 | 2 | 271 | ||
NM_026756.2 | NP_081032.2 | 2 | 271 | |||
GenBank mRNA | AK087539.1 | - | 2 | 256 | ||
AK147496.1 | - | 2 | 256 | |||
AK155258.1 | - | 2 | 273 | |||
U57635.1 | - | 2 | 211 | |||
Y07692.1 | - | 2 | 230 | |||
Y07693.1 | - | 2 | 576 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
