siRNA Details
Gene Symbol: Nefm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008691.2 | NP_032717.2 | 1 | 844 | ||
GenBank mRNA | AK051296.1 | - | 1 | 687 | ||
AK051696.1 | - | 1 | 684 | |||
AK080794.1 | - | 1 | 688 | |||
AK164318.1 | - | 1 | 687 | |||
AK165041.1 | - | 1 | 687 | |||
BC052352.1 | - | 1 | 686 | |||
BC058333.1 | - | 1 | 677 | |||
BC119602.2 | - | 1 | 652 | |||
BC128564.1 | - | 1 | 647 | |||
DQ201636.1 | - | 1 | 662 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_017029, XM_008770736, Z12152, M18628.
Gene Ontology Categories:
