siRNA Details
Gene Symbol: Mycn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008709.3 | NP_032735.3 | 3 | 1553 | ||
GenBank mRNA | AK144126.1 | - | 3 | 1554 | ||
AK146696.1 | - | 3 | 1554 | |||
AK146913.1 | - | 3 | 1551 | |||
BC005453.1 | - | 3 | 1437 | 3495446 | ||
BC049783.1 | - | 2 | 1482 | 6517457 | ||
M36277.1 | - | 1329 |
