siRNA Details
Gene Symbol: Map1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008634.2 | NP_032660.2 | 7 | 11419 | ||
GenBank mRNA | AK051573.1 | - | 1 | 2530 | ||
AK141735.1 | - | 1 | 2529 | |||
BC020992.1 | - | 1 | 414 | |||
BC037159.1 | - | 1 | 171 | |||
BC045135.1 | - | 1 | 1280 | |||
BC052015.1 | - | 1 | 510 | |||
BC057015.1 | - | 1 | 517 | |||
BC089345.1 | - | 1 | 221 | |||
BC094335.1 | - | 1 | 674 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
