siRNA Details
Gene Symbol: Pcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162946.1 | NP_001156418.1 | 4 | 385 | ||
NM_008797.3 | NP_032823.2 | 3 | 362 | |||
GenBank mRNA | AK128927.1 | - | 312 | |||
AK148328.1 | - | 4 | 332 | |||
AK170596.1 | - | 4 | 345 | |||
AK170871.1 | - | 4 | 335 | |||
AK172320.1 | - | 4 | 313 | |||
BC055030.1 | - | 4 | 320 | 5053475 | ||
BG087228.2 | - | 4 | 321 | |||
BY139237.1 | - | 3 | 360 | |||
L09192.1 | - | 3 | 338 |
