siRNA Details
Gene Symbol: Pdgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008808.3 | NP_032834.1 | 3 | 358 | ||
GenBank mRNA | AY324648.1 | - | 3 | 347 | ||
M29464.1 | - | 3 | 238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
