siRNA Details
Gene Symbol: Pira6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289428.1 | NP_001276357.1 | 1480 | |||
NM_008848.1 | NP_032874.1 | 1415 | ||||
GenBank mRNA | AF041035.1 | - | 1415 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
