siRNA Details
Gene Symbol: CALM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005184.2 | NP_005175.2 | 3 | 338 | ||
GenBank mRNA | AK094964.1 | - | 1 | 359 | ||
AK295927.1 | - | 3 | 215 | |||
AK310762.1 | - | 4 | 240 | |||
BC005137.2 | - | 3 | 242 | 3504793 | ||
BC006182.1 | - | 3 | 443 | 3528814 | ||
BG753029.1 | - | 3 | 339 | |||
BT006855.1 | - | 2 | 158 | |||
FJ224310.1 | - | 3 | 261 | |||
J04046.1 | - | 3 | 261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
