siRNA Details
Gene Symbol: Pbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008783.3 | NP_032809.1 | 8 | 2110 | ||
GenBank mRNA | AK087587.1 | - | 7 | 1234 | ||
BC054376.1 | - | 8 | 2071 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
