siRNA Details
Gene Symbol: Ptx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008987.3 | NP_033013.3 | 2, 3 | 715 | ||
GenBank mRNA | BQ442682.1 | - | 2, 3 | 602 | ||
X83601.1 | - | 2, 3 | 639 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
