siRNA Details
Gene Symbol: Psma2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008944.2 | NP_032970.2 | 3 | 233 | ||
GenBank mRNA | AF099733.1 | - | 3 | 200 | ||
AK012119.1 | - | 3 | 215 | |||
AK012990.1 | - | 3 | 222 | |||
AK035578.1 | - | 222 | ||||
AK136115.1 | - | 3 | 235 | |||
AK151883.1 | - | 3 | 224 | |||
AK152862.1 | - | 3 | 224 | |||
AK154011.1 | - | 3 | 217 | |||
AK167752.1 | - | 3 | 224 | |||
BC026768.1 | - | 3 | 206 | 3156907 | ||
CJ052982.1 | - | 3 | 235 | |||
X70303.1 | - | 3 | 277 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
