siRNA Details
Gene Symbol: OR2B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033057.2 | NP_149046.2 | 1 | 973 | ||
GenBank mRNA | BC136881.1 | - | 1 | 973 | 9020872 | |
BC136901.1 | - | 1 | 973 | 9020892 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
