siRNA Details
Gene Symbol: OR51B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033179.2 | NP_149419.2 | 1 | 453 | ||
GenBank mRNA | BC069094.1 | - | 1 | 453 | 7216908 | |
BC100919.1 | - | 1 | 452 | |||
BC148186.1 | - | 1 | 453 | 40025615 | ||
BC153815.1 | - | 1 | 453 | 40003856 |
