siRNA Details
Gene Symbol: Pola2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008893.3 | NP_032919.2 | 18 | 2363 | ||
GenBank mRNA | AK020656.1 | - | 4 | 625 | ||
AK133832.1 | - | 18 | 2349 | |||
AK150099.1 | - | 18 | 2305 | |||
AV373556.1 | - | 1 | 198 | |||
BC064795.1 | - | 18 | 2279 | 30111287 | ||
D13546.1 | - | 18 | 2296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
