siRNA Details
Gene Symbol: Rad23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009011.4 | NP_033037.2 | 2 | 400 | ||
GenBank mRNA | AK084917.1 | - | 2 | 392 | ||
AK142280.1 | - | 2 | 392 | |||
AK150089.1 | - | 2 | 392 | |||
AK157252.1 | - | 2 | 362 | |||
AK160880.1 | - | 2 | 392 | |||
AK160890.1 | - | 2 | 378 | |||
AK160973.1 | - | 2 | 378 | |||
AK167582.1 | - | 2 | 388 | |||
AK214312.1 | - | 2 | 93 | |||
BC027747.1 | - | 2 | 389 | 4459657 | ||
BC068193.1 | - | 2 | 359 | 6813997 | ||
X92411.1 | - | 2 | 80 |

More Information
Other Species Targeted

Gene Ontology Categories:
