siRNA Details
Gene Symbol: Rag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009019.2 | NP_033045.2 | 2 | 274 | ||
GenBank mRNA | AK088725.1 | - | 2 | 275 | ||
AK162974.1 | - | 2 | 274 | |||
BC138342.1 | - | 2 | 199 | 8861362 | ||
M29475.1 | - | 2 | 243 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
