siRNA Details
Gene Symbol: Ring1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009066.3 | NP_033092.3 | 3 | 417 | ||
GenBank mRNA | AK154962.1 | - | 3 | 378 | ||
AK155509.1 | - | 3 | 393 | |||
BC082771.1 | - | 1 | 595 | |||
BU614216.1 | - | 3 | 337 | |||
CV560731.1 | - | 1 | 50 | |||
KJ650216.1 | - | 2 | 194 | |||
Y12881.1 | - | 1 | 113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
