siRNA Details
Gene Symbol: MAP3K8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244134.1 | NP_001231063.1 | 2 | 596 | ||
NM_001320961.1 | NP_001307890.1 | 2 | 352 | |||
NM_005204.3 | NP_005195.2 | 3 | 827 | |||
XM_017015707.1 | XP_016871196.1 | 2 | 1044 | |||
XM_017015708.1 | XP_016871197.1 | 3 | 557 | |||
XM_017015709.1 | XP_016871198.1 | 2 | 3337 | |||
XM_017015710.1 | XP_016871199.1 | 2 | 447 | |||
XM_017015711.1 | XP_016871200.1 | 2 | 1044 | |||
XM_017015712.1 | XP_016871201.1 | 3 | 557 | |||
XM_017015713.1 | XP_016871202.1 | 3 | 801 | |||
XM_017015714.1 | XP_016871203.1 | 2 | 596 | |||
GenBank mRNA | AB209539.1 | - | 1 | 2958 | ||
BC104833.1 | - | 2 | 453 | 8143836 | ||
BC113566.1 | - | 2 | 453 | 8322618 | ||
BI753824.1 | - | 2 | 322 | |||
CR542284.1 | - | 1 | 215 | |||
D14497.1 | - | 3 | 581 | |||
Z14138.1 | - | 1 | 375 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
