siRNA Details
Gene Symbol: Rpl28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009081.2 | NP_033107.1 | 4 | 286 | ||
GenBank mRNA | AK012368.1 | - | 4 | 287 | ||
AK018714.1 | - | 4 | 287 | |||
AK076133.1 | - | 4 | 287 | |||
BC024395.1 | - | 4 | 286 | 4981342 | ||
BC086797.1 | - | 4 | 272 | 5713507 | ||
BC086932.1 | - | 4 | 288 | 6760499 | ||
BC092012.1 | - | 4 | 286 | 30608657 | ||
X74856.1 | - | 4 | 314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
