siRNA Details
Gene Symbol: Polr1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009087.2 | NP_033113.1 | 2 | 398 | ||
GenBank mRNA | AK002811.1 | - | 2 | 166 | ||
BM948930.1 | - | 2 | 695 | |||
D86609.1 | - | 2 | 203 |

More Information
Other Species Targeted

Gene Ontology Categories:
