siRNA Details
Gene Symbol: Rfx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009055.4 | NP_033081.3 | 21 | 3001 | ||
GenBank mRNA | AK037226.1 | - | 14 | 1944 | ||
AK083571.1 | - | 21 | 2972 | |||
AK147790.1 | - | 21 | 3042 | |||
BC057018.1 | - | 21 | 2980 | 5705637 | ||
X76088.1 | - | 21 | 2908 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
