siRNA Details
Gene Symbol: Rps17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009092.3 | NP_033118.1 | 3 | 260 | ||
GenBank mRNA | AK007992.1 | - | 3 | 268 | ||
AK010647.1 | - | 3 | 262 | |||
AK012781.1 | - | 3 | 249 | |||
AK158902.1 | - | 2 | 326 | |||
AK162022.1 | - | 3 | 254 | |||
AK162335.1 | - | 3 | 252 | |||
AK167206.1 | - | 3 | 254 | |||
BC081466.1 | - | 2 | 247 | 6412006 | ||
BC086900.1 | - | 3 | 253 | 6760363 | ||
BC086901.1 | - | 3 | 253 | 6763239 | ||
D25213.1 | - | 2 | 241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
