siRNA Details
Gene Symbol: Trim30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009099.2 | NP_033125.2 | 2 | 472 | ||
GenBank mRNA | AF220014.1 | - | 2 | 389 | ||
AF220015.1 | - | 2 | 389 | |||
AK089726.1 | - | 2 | 469 | |||
AK134144.1 | - | 2 | 441 | |||
AK137506.1 | - | 2 | 473 | |||
AK138249.1 | - | 2 | 475 | |||
BC005447.1 | - | 2 | 406 | 3493256 | ||
J03776.1 | - | 2 | 389 |
Gene Symbol: Trim30d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199146.2 | NP_954597.2 | 2 | 479 | ||
GenBank mRNA | BC056631.1 | - | 2 | 455 | 5035734 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
