siRNA Details
Gene Symbol: Rad54l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122958.1 | NP_001116430.1 | 2741 | |||
NM_001122959.1 | NP_001116431.1 | 2720 | ||||
NM_009015.3 | NP_033041.3 | 3071 | ||||
GenBank mRNA | X97796.1 | - | 2530 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
