siRNA Details
Gene Symbol: GNG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017998.3 | NP_001017998.1 | 3 | 305 | ||
NM_001198664.1 | NP_001185593.1 | 3 | 302 | |||
GenBank mRNA | BC010384.1 | - | 3 | 264 | ||
BC015206.2 | - | 3 | 251 | |||
BC016319.1 | - | 3 | 260 | |||
BC072671.1 | - | 3 | 253 | 5768882 | ||
BC107574.1 | - | 3 | 272 | 4050245 | ||
BG708408.1 | - | 3 | 310 | |||
DB504443.1 | - | 3 | 305 | |||
U31383.1 | - | 3 | 264 |
Gene Symbol: DNAJC25-GNG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004125.3 | NP_004116.2 | 3 | 549 | ||
GenBank mRNA | BC015391.1 | - | 3 | 515 | 4425229 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
