siRNA Details
Gene Symbol: Rsu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009105.4 | NP_033131.2 | 4 | 425 | ||
GenBank mRNA | AK009288.1 | - | 4 | 318 | ||
AK011857.1 | - | 4 | 318 | |||
AK135491.1 | - | 5 | 408 | |||
AK149835.1 | - | 4 | 295 | |||
AK150011.1 | - | 4 | 293 | |||
AK152655.1 | - | 4 | 305 | |||
AK169076.1 | - | 4 | 343 | |||
BC003827.1 | - | 4 | 288 | 3493122 | ||
CX228720.1 | - | 4 | 425 | |||
X63039.1 | - | 4 | 240 |
