siRNA Details
Gene Symbol: Satb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163630.1 | NP_001157102.1 | 2 | 488 | ||
NM_001163631.1 | NP_001157103.1 | 2 | 356 | |||
NM_001163632.1 | NP_001157104.1 | 2 | 329 | |||
NM_009122.2 | NP_033148.2 | 2 | 379 | |||
GenBank mRNA | AK037740.1 | - | 2 | 379 | ||
AK040502.1 | - | 1 | 1835 | |||
AK088563.1 | - | 2 | 355 | |||
AK134040.1 | - | 2 | 359 | |||
AK158518.1 | - | 2 | 489 | |||
BB635001.1 | - | 2 | 329 | |||
BC011132.1 | - | 2 | 239 | 4164993 | ||
CT010371.1 | - | 1 | 167 | |||
U05252.1 | - | 2 | 365 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001012129, XM_008766763, XM_006244219, XM_006244221, XM_006244220, and others.
Gene Ontology Categories:
