siRNA Details
Gene Symbol: Serpinb3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009126.3 | NP_033152.3 | 3 | 341 | ||
GenBank mRNA | AY144683.1 | - | 3 | 325 | ||
AY144684.1 | - | 3 | 325 | |||
BC063756.1 | - | 3 | 341 | 30288465 |
Gene Symbol: Serpinb3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198680.2 | NP_941373.1 | 4 | 365 | ||
GenBank mRNA | AK003220.1 | - | 4 | 366 | ||
AY144685.1 | - | 3 | 325 | |||
AY144686.1 | - | 3 | 325 | |||
AY367774.1 | - | 3 | 309 | |||
BC099584.1 | - | 4 | 362 | 30291007 |
Gene Symbol: Serpinb3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201363.2 | NP_958751.2 | 4 | 375 | ||
GenBank mRNA | AK003650.1 | - | 4 | 376 | ||
AY367775.1 | - | 3 | 305 | |||
BC132091.1 | - | 3 | 327 | 40130368 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
