siRNA Details
Gene Symbol: ETV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145312.1 | NP_001138784.1 | 3 | 438 | ||
NM_005240.1 | NP_005231.1 | 3 | 236 | |||
XM_006711210.2 | XP_006711273.1 | 3 | 419 | |||
GenBank mRNA | AF218540.1 | - | 2 | 149 | ||
AK304789.1 | - | 3 | 242 | |||
AK316274.1 | - | 3 | 438 | |||
BC022868.1 | - | 3 | 236 | |||
L16464.1 | - | 3 | 215 |
Gene Symbol: ETV3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004341.2 | NP_001004341.1 | 2 | 446 | ||
GenBank mRNA | AK131392.1 | - | 2 | 446 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_012051, NM_001083318, NM_001286844, AF218539, BC057151, and others.
Gene Ontology Categories:
