siRNA Details
Gene Symbol: MAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031804.2 | NP_001026974.1 | 1 | 1866 | ||
NM_005360.4 | NP_005351.2 | 1 | 1866 | |||
XM_017023233.1 | XP_016878722.1 | 1 | 1550 | |||
XM_017023234.1 | XP_016878723.1 | 1 | 1550 | |||
XM_017023235.1 | XP_016878724.1 | 1 | 1550 | |||
GenBank mRNA | AF055376.1 | - | 1 | 1850 | ||
AF055377.1 | - | 1 | 1850 | |||
BC081542.1 | - | 1 | 1158 | 30347784 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
