siRNA Details
Gene Symbol: Zfp106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089359.1 | - | 5 | 2704 | ||
AK155019.1 | - | 5 | 2704 | |||
U59739.1 | - | 2 | 2774 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
