siRNA Details
Gene Symbol: EGFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005228.3 | NP_005219.2 | 28 | 5175 | ||
GenBank mRNA | HQ912715.1 | - | 27 | 5046 | ||
X00588.1 | - | 28 | 5115 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
