siRNA Details
Gene Symbol: Serpina1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009245.2 | NP_033271.1 | 3 | 980 | ||
GenBank mRNA | M75720.1 | - | 3 | 917 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
