siRNA Details
Gene Symbol: Src | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009271.3 | NP_033297.2 | 5, 7 | 742 | ||
GenBank mRNA | M17031.1 | - | 2, 4 | 358 |

More Information
Other Species Targeted

Gene Ontology Categories:
