siRNA Details
Gene Symbol: Sms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009214.3 | NP_033240.3 | 2 | 230 | ||
GenBank mRNA | AF031486.1 | - | 2 | 213 | ||
AK002687.1 | - | 2 | 228 | |||
AK011542.1 | - | 2 | 232 | |||
AK017775.1 | - | 2 | 201 | |||
AK049780.1 | - | 2 | 229 | |||
AK049787.1 | - | 2 | 219 | |||
AK136798.1 | - | 2 | 231 | |||
AK149929.1 | - | 2 | 228 | |||
AK160637.1 | - | 2 | 225 | |||
AK160659.1 | - | 2 | 231 | |||
AK166665.1 | - | 2 | 256 | |||
BC046623.1 | - | 2 | 212 | 6315420 | ||
BC058688.1 | - | 2 | 471 | 6413869 | ||
Y09419.1 | - | 2 | 203 |
