siRNA Details
Gene Symbol: Src | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009271.3 | NP_033297.2 | 6, 7 | 746 | ||
GenBank mRNA | M17031.1 | - | 3, 4 | 362 |

More Information
Other Species Targeted

Gene Ontology Categories:
