siRNA Details
Gene Symbol: Ssr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166480.1 | NP_001159952.1 | 4 | 529 | ||
NM_009279.3 | NP_033305.1 | 4 | 411 | |||
GenBank mRNA | AK003044.1 | - | 4 | 391 | ||
AK007921.1 | - | 4 | 367 | |||
AK013723.1 | - | 4 | 373 | |||
AK160089.1 | - | 4 | 377 | |||
AK191259.1 | - | 2 | 111 | |||
BC008172.1 | - | 4 | 332 | 3481948 | ||
BC012971.1 | - | 2 | 2068 | |||
BC019419.1 | - | 3 | 1862 | |||
BI989805.1 | - | 4 | 529 | |||
BQ747160.1 | - | 4 | 314 | |||
CJ144362.1 | - | 4 | 416 | |||
X90582.1 | - | 4 | 492 |
